Formaldehyde sodium bisulfite 95%

Supplier: Thermo Scientific Chemicals

A16194-0E A16194-22 A16194-36
CAAAA16194-0EEA 390.26 CAD
CAAAA16194-0E CAAAA16194-22 CAAAA16194-36
Formaldehyde sodium bisulfite 95%
Formaldehyde sodium bisulfite

Purity: 95% CAS No.: 870-72-4 Molecular Formula: HOCH2SO3Na Formula Weight: 134.09 Odor: Odorless Melting Point: 200° dec. MDL No.: MFCD00044664

Formula: CH₄O₄S
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044664
CAS Number: 870-72-4
EINECS: 212-800-9
Merck Index: 14,4236

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR