3-Hydroxy-2-naphthoic acid hydrazide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Hydroxy-2-naphthohydrazide, 2-Hydroxy-3-naphthoic Acid Hydrazide

A16192-14 A16192-22
CAAAA16192-14EA 87.53 CAD
CAAAA16192-14 CAAAA16192-22
3-Hydroxy-2-naphthoic acid hydrazide 98%
3-Hydroxy-2-naphthoic acid hydrazide

Purity: 98% CAS No.: 5341-58-2 Molecular Formula: C11H10N2O2 Formula Weight: 202.21 Melting Point: ca 205° dec. Beilstein Reference: 395153 MDL No.: MFCD00004097

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Melting Pt: ca 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004097
CAS Number: 5341-58-2
EINECS: 226-282-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR