DL-Histidine monohydrochloride monohydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-His-OH.HCl.H2O, H-DL-His-OH·HCl·H₂O

A16165-06 A16165-14 A16165-22
CAAAA16165-06EA 38.24 CAD
CAAAA16165-06 CAAAA16165-14 CAAAA16165-22
DL-Histidine monohydrochloride monohydrate 99%
DL-Histidine monohydrochloride monohydrate

Purity: 99% CAS No.: 123333-71-1 Molecular Formula: C6H9N3O2.HCl.H2O Formula Weight: 209.63 (191.62anhy) Melting Point: ca 250° dec. MDL No.: MFCD00151028

Formula: C₆H₉N₃O₂.HCl.H₂O
Melting Pt: ca 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151028
CAS Number: 123333-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR