Chlorophyllin, coppered trisodium salt

Supplier: Thermo Scientific Chemicals

A16155-06 A16155-14 A16155-22
CAAAA16155-06EA 42.82 CAD
CAAAA16155-06 CAAAA16155-14 CAAAA16155-22
Chlorophyllin, coppered trisodium salt
Chlorophyllin, coppered trisodium salt

CAS No.: 11006-34-1 Molecular Formula: C34H31CuN4Na3O6 Formula Weight: 724.15 Storage Temperature: 2°C to 8°C Odor: Odorless MDL No.: MFCD00012149

Formula: C₃₄H₃₁CuN₄Na₃O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00012149
CAS Number: 11006-34-1
EINECS: 234-242-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR