Strontium acetate hemihydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Strontium Acetate Hemihydrate

A16149-0E A16149-22 A16149-36
CAAAAA16149-0EEA 894.28 CAD
CAAAAA16149-0E CAAAAA16149-22 CAAAAA16149-36
Strontium acetate hemihydrate 98%
Strontium acetate hemihydrate

Purity: 98% CAS No.: 14692-29-6 Molecular Formula: Sr(OOCCH3)2.0.5H2O Formula Weight: 214.72 (205.71anhy) Melting Point: 150° -0.5H2O MDL No.: MFCD00198064

Formula: C₈H₁₄O₉Sr₂
MW: 429.43 g/mol
Melting Pt: 150 °C (-0.5H₂O) (–H₂O of crystall.)
Storage Temperature: Ambient
MDL Number: MFCD00198064
CAS Number: 14692-29-6
EINECS: 208-854-8
Merck Index: 14,8836

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR