(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16140-06
A16140-14
CAAAA16140-06EA
117.24
CAD
CAAAA16140-06
CAAAA16140-14
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) 98%
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
Fieser: 6,34 7,14 18,35 20,22
Purity: 98%
CAS No.: 56602-33-6
Molecular Formula: C12H22F6N6OP2
Formula Weight: 442.29
Storage Temperature: 2°C to 8°C
Melting Point: ca 138° dec.
Beilstein Reference: 4287025
UN No.: 1325
MDL No.: MFCD00011948
Formula:
C₁₂H₂₂F₆N₆OP₂ Melting Pt: ca 138 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011948 CAS Number: 56602-33-6 EINECS: 260-279-1 UN: 1325 ADR: "4.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...