4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 18-Nitro-2,3,5,6,8,9,11,12,14,15-decahydrobenzo[b][1,4,7,10,13,16]hexaoxacyclooctadecine

A16138-03 A16138-06
CAAAA16138-03EA 140.62 CAD
CAAAA16138-03 CAAAA16138-06
4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) 99%
4'-Nitrobenzo-18-crown-6 (4'-Nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane)

Purity: 99% CAS No.: 53408-96-1 Molecular Formula: C16H23NO8 Formula Weight: 357.35 Melting Point: 84-86° MDL No.: MFCD00143207

Formula: C₁₆H₂₃NO₈
MW: 357.35 g/mol
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00143207
CAS Number: 53408-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR