3-Bromobenzylamine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16132-06
A16132-14
CAAAA16132-06EA
61.62
CAD
CAAAA16132-06
CAAAA16132-14
CAAAA16132-22
3-Bromobenzylamine hydrochloride 98%
3-Bromobenzylamine hydrochloride
Purity: 98%
CAS No.: 39959-54-1
Molecular Formula: C7H8BrN.HCl
Formula Weight: 222.52
Melting Point: 218-221°
Beilstein Reference: 3913185
Formula:
C₇H₉BrClN Melting Pt: 218…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012856 CAS Number: 39959-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...