α-Isopropylphenylacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16077-14
A16077-06
CAAAAA16077-14EA
635.16
CAD
CAAAAA16077-14
CAAAA16077-03
CAAAA16077-06
α-Isopropylphenylacetic acid 97%
alpha-Isopropylphenylacetic acid
Purity: 97%
CAS No.: 3508-94-9
Molecular Formula: C11H14O2
Formula Weight: 178.23
Melting Point: 57-62°
Beilstein Reference: 2045915
MDL No.: MFCD00021708
Formula:
C₁₁H₁₄O₂ Melting Pt: 57…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021708 CAS Number: 3508-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...