Sodium percarbonate 13 - 14% (active oxygen)
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16045-0E
A16045-22
A16045-36
CAAAAA16045-0EEA
178.54
CAD
CAAAAA16045-0E
CAAAAA16045-22
CAAAAA16045-36
Sodium percarbonate 13 - 14% (active oxygen)
Sodium percarbonate
Fieser: 19,314 21,405
CAS No.: 15630-89-4
Molecular Formula: Na2CO3.1.5H2O2
Formula Weight: 157.01
UN No.: 1479
MDL No.: MFCD00043204
Formula:
2Na₂CO₃·3H₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00043204 CAS Number: 15630-89-4 EINECS: 239-707-6 UN: 3378 ADR: "5.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...