6-Methylnicotinamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16039-03
A16039-06
CAAAA16039-03EA
77.26
CAD
CAAAA16039-03
CAAAA16039-06
6-Methylnicotinamide 98%
6-Methylnicotinamide
Purity: 98%
CAS No.: 6960-22-1
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Melting Point: 195-199°
Beilstein Reference: 112050
Formula:
C₇H₈N₂O Melting Pt: 195…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006342 CAS Number: 6960-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...