Nε-carbobenzoxy-L-lysine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nε-Cbz-L-lysine, (S)-2-Amino-6-(((benzyloxy)carbonyl)amino)hexanoic acid, H-Lys(Z)-OH
A16022-06
A16022-14
CAAAA16022-03EA
27.87
CAD
CAAAA16022-03
CAAAA16022-06
CAAAA16022-14
Nε-carbobenzoxy-L-lysine 98%
Nepsilon-carbobenzoxy-L-lysine
Purity: 98%
CAS No.: 1155-64-2
Molecular Formula: C14H20N2O4
Formula Weight: 280.32
Melting Point: 259° dec.
Beilstein Reference: 2222482
MDL No.: MFCD00002638
Formula:
C₁₄H₂₀N₂O₄ MW: 280.32 g/mol Melting Pt: 259 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002638 CAS Number: 1155-64-2 EINECS: 214-585-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...