Boc-L-isoleucine ≥98%
Supplier: Thermo Scientific Chemicals
A16021-06
A16021-14
CAAAA16021-03EA
37.94
CAD
CAAAA16021-03
CAAAA16021-06
CAAAA16021-14
Boc-L-isoleucine ≥98%
Boc-L-isoleucine
Purity: 98+%
CAS No.: 13139-16-7
Molecular Formula: C11H21NO4
Formula Weight: 231.06
Storage Temperature: 2°C to 8°C
Melting Point: 67-70°
Beilstein Reference: 1711700
MDL No.: MFCD00038324
Formula:
C₁₁H₂₁NO₄ Melting Pt: 67…70 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038324 CAS Number: 13139-16-7 EINECS: 236-074-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...