N-ɑ-Boc-L-asparagine ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16019-06
A16019-14
CAAAA16019-06EA
40.13
CAD
CAAAA16019-06
CAAAA16019-14
N-ɑ-Boc-L-asparagine ≥98%
N-alpha-Boc-L-asparagine
Purity: 98+%
CAS No.: 7536-55-2
Molecular Formula: C9H16N2O5
Formula Weight: 232.24
Storage Temperature: 2°C to 8°C
Melting Point: ca 170° dec.
Beilstein Reference: 1977963
Optical Rotation: -7.3° (c=2 in DMF)
MDL No.: MFCD00038152
Formula:
C9H16N2O5 Melting Pt: ca 170 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038152 CAS Number: 7536-55-2 EINECS: 231-405-2 |
Specification Test Results
Optical rotation | -7.3° (c=2 in DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...