N-Boc-L-alanine ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH

A16018-06 A16018-14
CAAAA16018-06EA 51.82 CAD
CAAAA16018-06 CAAAA16018-14
N-Boc-L-alanine ≥98%
N-Boc-L-alanine

Purity: 98+% CAS No.: 15761-38-3 Molecular Formula: C8H15NO4 Formula Weight: 189.21 Storage Temperature: 2°C to 8°C Melting Point: 80-84° Beilstein Reference: 1726365 Optical Rotation: -26° (c=2 in acetic acid) MDL No.: MFCD00037225

Formula: C8H15NO4
Melting Pt: 80…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037225
CAS Number: 15761-38-3
EINECS: 239-847-8

Order Now

Specification Test Results

Optical rotation -26° (c=2 in acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR