N-(tert-Butoxycarbonyl)-L-phenylalanine 99%
Supplier: Thermo Scientific Chemicals
A16017-06
A16017-14
CAAAA16017-06EA
54.83
CAD
CAAAA16017-06
CAAAA16017-14
N-(tert-Butoxycarbonyl)-L-phenylalanine 99%
N-(tert-Butoxycarbonyl)-L-phenylalanine
Purity: 99%
CAS No.: 13734-34-4
Molecular Formula: C14H19NO4
Formula Weight: 265.31
Storage Temperature: 2°C to 8°C
Melting Point: 84-88°
Beilstein Reference: 2219729
Optical Rotation: +25° (c=1 in ethanol)
MDL No.: MFCD00002663
Formula:
C₁₄H₁₉NO₄ Melting Pt: 84…88 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002663 CAS Number: 13734-34-4 EINECS: 237-305-5 |
Specification Test Results
Optical rotation | +25° (c=1 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...