N-Acetonylpyridinium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16015-06
A16015-14
CAAAA16015-06EA
89.74
CAD
CAAAA16015-06
CAAAA16015-14
CAAAA16015-22
N-Acetonylpyridinium chloride 97%
N-Acetonylpyridinium chloride
Purity: 97%
CAS No.: 42508-60-1
Molecular Formula: C8H10ClNO
Formula Weight: 171.63
MDL No.: MFCD00031949
Formula:
C₈H₁₀ClNO Storage Temperature: Ambient |
MDL Number:
MFCD00031949 CAS Number: 42508-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...