Tetrazolium violet 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16008-03
A16008-06
A16008-14
CAAAA16008-03EA
72.05
CAD
CAAAA16008-03
CAAAA16008-06
CAAAA16008-14
Tetrazolium violet 98+%
Tetrazolium Violet
Purity: 98+%
CAS No.: 1719-71-7
Molecular Formula: C23H17ClN4
Formula Weight: 384.87
Melting Point: 247-249°
MDL No.: MFCD00011875
Formula:
C₂₃H₁₇ClN₄ Melting Pt: 247…249 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011875 CAS Number: 1719-71-7 EINECS: 217-008-7 UN: 1325 ADR: "4.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...