N-Boc-L-valine 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Val-OH, BOC-L-Valine

A16007-06 A16007-14
CAAAA16007-06EA 67.31 CAD
CAAAA16007-06 CAAAA16007-14
N-Boc-L-valine 98+%
N-Boc-L-valine

Purity: 98+% CAS No.: 13734-41-3 Molecular Formula: C10H19NO4 Formula Weight: 217.27 Storage Temperature: 2°C to 8°C Melting Point: 77-80° Beilstein Reference: 1711290 Optical Rotation: -7.5° (c=1 in acetic acid) MDL No.: MFCD00065605

Formula: C₁₀H₁₉NO₄
Melting Pt: 77…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065605
CAS Number: 13734-41-3
EINECS: 237-307-6

Order Now

Specification Test Results

Optical rotation -7.5° (c=1 in acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR