Theophyllin monohydrate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15997-0B
A15997-18
A15997-30
CAAAA15997-0BEA
425.02
CAD
CAAAA15997-0B
CAAAA15997-18
CAAAA15997-30
Theophyllin monohydrate 99%
Theophyllin monohydrate
Dimethylxanthine. A methylxanthine drug used in therapy for respiratory diseases such as COPD and asthma
Purity: 99%
CAS No.: 5967-84-0
Molecular Formula: C7H8N4O2.H2O
Formula Weight: 198.18 (180.16anhy)
Odor: Odorless
Melting Point: 270-274°
Beilstein Reference: 13463
Merck Reference: 14,9285
UN No.: 2811
MDL No.: MFCD00151659
Formula:
C₇H₈N₄O₂·1H₂O Melting Pt: 270…274 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151659 CAS Number: 5967-84-0 EINECS: 200-385-7 UN: 2811 ADR: "6.1",III Merck Index: 14,9285 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...