4,5-Dibromo-3(2H)-pyridazinone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15994-06
A15994-14
CAAAA15994-06EA
213.3
CAD
CAAAA15994-06
CAAAA15994-14
4,5-Dibromo-3(2H)-pyridazinone 98%
4,5-Dibromo-3(2H)-pyridazinone
Purity: 98%
CAS No.: 5788-58-9
Molecular Formula: C4H2Br2N2O
Formula Weight: 253.89
Melting Point: ca 220° dec.
Beilstein Reference: 121469
Formula:
C₄H₂Br₂N₂O Melting Pt: ca 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00023641 CAS Number: 5788-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...