1H-Benzimidazole-5-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Benzo[d]imidazole-5-carboxylic acid, Benzimidazole-5-carboxylic acid, 5-Benzimidazolecarboxylic acid

A15980-14 A15980-09 A15980-18 A15980-30
CAAAAA15980-14EA 279.66 CAD
CAAAAA15980-14 CAAAA15980-09 CAAAA15980-18 CAAAA15980-30
1H-Benzimidazole-5-carboxylic acid 98%
1H-Benzimidazole-5-carboxylic acid

Purity: 98% CAS No.: 15788-16-6 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 300° Beilstein Reference: 5686

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011555
CAS Number: 15788-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR