2,4,6-Trimethylstyrene 95% stabilized

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3,5-Trimethyl-2-vinylbenzene, 2-Vinylmesitylene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC) 500 ppm

A15974-03 A15974-06 A15974-14
CAAAAA15974-03EA 46.93 CAD
CAAAAA15974-03 CAAAA15974-06 CAAAA15974-14
2,4,6-Trimethylstyrene 95% stabilized
2,4,6-Trimethylstyrene

Stab. with 500ppm 4-tert-butylcatechol


Purity: 95% CAS No.: 769-25-5 Molecular Formula: C11H14 Formula Weight: 146.23 Storage Temperature: 2°C to 8°C Flash Point: 75°(167°F) Refractive Index: 1.5320 Boiling Point: 208-209° Beilstein Reference: 1904365 MDL No.: MFCD00008613

Formula: C₁₁H₁₄
MW: 146.23 g/mol
Boiling Pt: 208…209 °C
Density: 0.908
Flash Pt: 75 °C(167 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008613
CAS Number: 769-25-5
EINECS: 212-205-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR