3-(4-Methoxyphenyl)-3-oxopropanenitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A15973-06 A15973-14
CAAAA15973-06EA 183.28 CAD
CAAAA15973-06 CAAAA15973-14
3-(4-Methoxyphenyl)-3-oxopropanenitrile 98%
3-(4-Methoxyphenyl)-3-oxopropanenitrile

Purity: 98% CAS No.: 3672-47-7 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 125-130° Beilstein Reference: 743909 MDL No.: MFCD00067889

Formula: C₁₀H₉NO₂
Melting Pt: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00067889
CAS Number: 3672-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR