Benzyl-4-chlorophenyl ketone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Benzyl 4-chlorophenyl ketone, 4'-Chloro-2-phenylacetophenone
A15959-06
A15959-14
A15959-22
CAAAA15959-06EA
77.42
CAD
CAAAA15959-06
CAAAA15959-14
CAAAA15959-22
Benzyl-4-chlorophenyl ketone 98%
Benzyl 4-chlorophenyl ketone
Purity: 98%
CAS No.: 1889-71-0
Molecular Formula: C14H11ClO
Formula Weight: 230.69
Melting Point: 103-107°
Beilstein Reference: 976443
Formula:
C₁₄H₁₁ClO Melting Pt: 103…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016342 CAS Number: 1889-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...