(1S,2R)-(-)-10,2-Camphorsultam 99%

Supplier: Thermo Scientific Chemicals
Warning

A15897-06 A15897-14
CAAAA15897-03EA 226.02 CAD
CAAAA15897-03 CAAAA15897-06 CAAAA15897-14
(1S,2R)-(-)-10,2-Camphorsultam 99%
(1S,2R)-(-)-10,2-Camphorsultam

Fieser: 13,62 14,71 17,65


Purity: 99% CAS No.: 94594-90-8 Molecular Formula: C10H17NO2S Formula Weight: 215.32 Melting Point: 181-186° Beilstein Reference: 83811 Optical Rotation: -33° (c=1 in ethanol) MDL No.: MFCD00066271

Formula: C10H17NO2S
Melting Pt: 181…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00066271
CAS Number: 94594-90-8

Order Now

Specification Test Results

Optical rotation -33° (c=1 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR