L-Valine methyl ester hydrochloride 99%
Supplier: Thermo Scientific Chemicals
A15892-09
A15892-18
CAAAA15892-09EA
104.91
CAD
CAAAA15892-09
CAAAA15892-18
L-Valine methyl ester hydrochloride 99%
L-Valine methyl ester hydrochloride
Purity: 99%
CAS No.: 6306-52-1
Molecular Formula: C6H13NO2.HCl
Formula Weight: 167.64
Storage Temperature: 2°C to 8°C
Melting Point: 166° dec.
Beilstein Reference: 3594960
Optical Rotation: +16° (c=2 in water)
MDL No.: MFCD00012497
Formula:
C₆H₁₄ClNO₂ Melting Pt: 166 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012497 CAS Number: 6306-52-1 EINECS: 228-620-9 |
Specification Test Results
Optical rotation | +16° (c=2 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...