Tetrahydro-4H-thiopyran-4-one 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrahydrothiopyran-4-one

A15872-03 A15872-06
CAAAA15872-03EA 66.36 CAD
CAAAA15872-03 CAAAA15872-06
Tetrahydro-4H-thiopyran-4-one 99%
Tetrahydro-4H-thiopyran-4-one

Purity: 99% CAS No.: 1072-72-6 Molecular Formula: C5H8OS Formula Weight: 116.18 Odor: Weak Melting Point: 60-64° MDL No.: MFCD00006660

Formula: C₅H₈OS
MW: 116.18 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00006660
CAS Number: 1072-72-6
EINECS: 214-015-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR