5-Amino-1-methyl-3-phenyl-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15754-03
A15754-06
CAAAA15754-03EA
36.02
CAD
CAAAA15754-03
CAAAA15754-06
CAAAA15754-14
5-Amino-1-methyl-3-phenyl-1H-pyrazole 97%
5-Amino-1-methyl-3-phenyl-1H-pyrazole
Purity: 97%
CAS No.: 10199-50-5
Molecular Formula: C10H11N3
Formula Weight: 173.22
Odor: Odorless
Melting Point: 124-130°
Beilstein Reference: 743807
MDL No.: MFCD00067874
Formula:
C₁₀H₁₁N₃ Melting Pt: 124…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067874 CAS Number: 10199-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...