5-Chloro-6-hydroxynicotinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Chloro-6-hydroxy-3-pyridinecarboxylic Acid
A15737-06
A15737-14
CAAAA15737-06EA
41.87
CAD
CAAAA15737-06
CAAAA15737-14
5-Chloro-6-hydroxynicotinic acid 98%
5-Chloro-6-hydroxynicotinic acid
Purity: 98%
CAS No.: 54127-63-8
Molecular Formula: C6H4ClNO3
Formula Weight: 173.56
Melting Point:
Formula:
C₆H₄ClNO₃ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052950 CAS Number: 54127-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...