Ellagic acid, (∼12% H₂O) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3,7,8-Tetrahydroxychromeno[5,4,3-cde]chromene-5,10-dione, 4,4',5,5',6,6'-Hexahydroxydiphenic acid 2,6,2',6'-dilactone
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.
Purity: 97%
CAS No.: 476-66-4
Molecular Formula: C14H6O8.xH2O
Formula Weight: 302.20(anhy)
Melting Point: 450° dec.
Beilstein Reference: 47549
MDL No.: MFCD00149494
Formula:
C₁₄H₆O₈ Melting Pt: 450 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006914 CAS Number: 476-66-4 EINECS: 207-508-3 Merck Index: 14,3547 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...