4-Benzyl-4-hydroxypiperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Benzylpiperidin-4-ol

A15664-03 A15664-06 A15664-14
CAAAAA15664-03EA 39.97 CAD
CAAAAA15664-03 CAAAAA15664-06 CAAAAA15664-14
4-Benzyl-4-hydroxypiperidine 97%
4-Benzyl-4-hydroxypiperidine

Purity: 97% CAS No.: 51135-96-7 Molecular Formula: C12H17NO Formula Weight: 191.27 Melting Point: ca 80° Beilstein Reference: 1451803

Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
Melting Pt: ∼ 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00023147
CAS Number: 51135-96-7
EINECS: 257-003-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR