Bis(trimethylsilyl)methane 98%

Supplier: Thermo Scientific Chemicals
Danger

A15662-06 A15662-14
CAAAA15662-03EA 148.01 CAD
CAAAA15662-03 CAAAA15662-06 CAAAA15662-14
Bis(trimethylsilyl)methane 98%
Bis(trimethylsilyl)methane

Purity: 98% CAS No.: 2117-28-4 Molecular Formula: (CH3)3SiCH2Si(CH3)3 Formula Weight: 160.41 Flash Point: 12°(53°F) Refractive Index: 1.4170 Melting Point: -71° Boiling Point: 132-133° Beilstein Reference: 1734530 UN No.: 1993 MDL No.: MFCD00008275

Formula: C₇H₂₀Si₂
MW: 160.405 g/mol
Boiling Pt: 132…133 °C
Melting Pt: –71 °C
Density: 0.750
Flash Pt: 12 °C (53 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008275
CAS Number: 2117-28-4
EINECS: 218-322-7
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR