(1-Ethoxycarbonylethylidene)triphenylphosphorane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(Carbethoxyethylidene)triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)propanoate
A15619-14
A15619-09
A15619-18
A15619-30
CAAAAA15619-14EA
295.46
CAD
CAAAAA15619-14
CAAAA15619-09
CAAAA15619-18
CAAAA15619-30
(1-Ethoxycarbonylethylidene)triphenylphosphorane 97%
(1-Ethoxycarbonylethylidene)triphenylphosphorane
Purity: 97%
CAS No.: 5717-37-3
Molecular Formula: C23H23O2P
Formula Weight: 362.41
Storage Temperature: 2°C to 8°C
Melting Point: 154-160°
Beilstein Reference: 757112
MDL No.: MFCD00009160
Formula:
C₂₃H₂₃O₂P MW: 362.41 g/mol Melting Pt: 154…160 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009160 CAS Number: 5717-37-3 EINECS: 227-215-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...