Ethyl malonyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15616-06
A15616-14
A15616-22
CAAAA15616-06EA
109.81
CAD
CAAAA15616-06
CAAAA15616-14
CAAAA15616-22
Ethyl malonyl chloride 95%
Ethyl malonyl chloride
Purity: 95%
CAS No.: 36239-09-5
Molecular Formula: C5H7ClO3
Formula Weight: 150.56
Storage Temperature: 2°C to 8°C
Flash Point: 67°(152°F)
Refractive Index: 1.4290
Boiling Point: 83°/22mm
Beilstein Reference: 636215
UN No.: 3265
MDL No.: MFCD00000736
Formula:
C₅H₇ClO₃ Boiling Pt: 83 °C/22 mm Density: 1.189 Flash Pt: 67 °C (152 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000736 CAS Number: 36239-09-5 EINECS: 252-934-5 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...