1-Acetylpiperidine-4-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15606-22 A15606-06 A15606-14
CAAAAA15606-22EA 355.5 CAD
CAAAAA15606-22 CAAAA15606-06 CAAAA15606-14
1-Acetylpiperidine-4-carboxylic acid 98+%
1-Acetylpiperidine-4-carboxylic acid

Purity: 98+% CAS No.: 25503-90-6 Molecular Formula: C8H13NO3 Formula Weight: 171.19 Melting Point: 180-184° Beilstein Reference: 389577

Formula: C₈H₁₃NO₃
MW: 171.19 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00023706
CAS Number: 25503-90-6
EINECS: 247-049-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR