Methyl-4-bromocrotonate tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15582-09
A15582-18
A15582-30
CAAAA15582-09EA
76.16
CAD
CAAAA15582-09
CAAAA15582-18
CAAAA15582-30
Methyl-4-bromocrotonate tech. 85%, Technical Grade
Methyl 4-bromocrotonate
Purity: 85%
CAS No.: 1117-71-1
Molecular Formula: C5H7BrO2
Formula Weight: 179.01
Storage Temperature: 2°C to 8°C
Flash Point: 91°(195°F)
Refractive Index: 1.5010
Boiling Point: 83-85°/13mm
Beilstein Reference: 1745755
UN No.: 1760
MDL No.: MFCD00000246
Formula:
C₅H₇BrO₂ Boiling Pt: 83…85 °C/13 mm Density: 1.524 Flash Pt: 91 °C(195 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000246 CAS Number: 1117-71-1 EINECS: 214-251-0 UN: 1760 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...