Aniline sulfate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15563-22
A15563-36
CAAAA15563-0EEA
1403.88
CAD
CAAAA15563-0E
CAAAA15563-22
CAAAA15563-36
Aniline sulfate 98%
Aniline sulphate
Soluble in water
Purity: 98%
CAS No.: 542-16-5
Molecular Formula: (C6H5NH2)2.H2SO4
Formula Weight: 284.34
Odor: Odorless
Solubility: Soluble in water
Beilstein Reference: 3729533
UN No.: 2811
MDL No.: MFCD00013110
Formula:
C₁₂H₁₆N₂O₄S Density: 1.377 Storage Temperature: Ambient |
MDL Number:
MFCD00013110 CAS Number: 542-16-5 EINECS: 208-805-0 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...