Homophthalic anhydride 97+%

Supplier: Thermo Scientific Chemicals
Warning

A15538-09 A15538-18
CAAAA15538-09EA 85.32 CAD
CAAAA15538-09 CAAAA15538-18 CAAAA15538-30
Homophthalic anhydride 97+%
Homophthalic anhydride

Fieser: 11,254


Purity: 97+% CAS No.: 703-59-3 Molecular Formula: C9H6O3 Formula Weight: 162.14 Melting Point: 140-144° Beilstein Reference: 5244

Formula: C₉H₆O₃
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00006894
CAS Number: 703-59-3
EINECS: 211-873-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR