Polyethylene oxide, M.W. >5,000,000
Supplier: Thermo Scientific Chemicals
A15536-22
A15536-36
CAAAA15536-22EA
178.54
CAD
CAAAA15536-22
CAAAA15536-36
Polyethylene oxide, M.W. >5,000,000
Polyethylene oxide
CAS No.: 25322-68-3
Molecular Formula: (-CH2CH2O-)n
Odor: Odorless
pH: 5-7
Melting Point: ca 175°
MDL No.: MFCD00147992
Formula:
(-CH₂CH₂O-)n Melting Pt: ∼ 175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00147970 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...