D-(+)-Glucosamine hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
A15532-0B
A15532-18
A15532-30
CAAAA15532-0BEA
597.24
CAD
CAAAA15532-0B
CAAAA15532-18
CAAAA15532-30
D-(+)-Glucosamine hydrochloride ≥98%
D-(+)-Glucosamine hydrochloride
Purity: 98+%
CAS No.: 66-84-2
Molecular Formula: C6H13NO5.HCl
Formula Weight: 215.64
Melting Point: ca 192° dec.
Beilstein Reference: 4157370
Optical Rotation: +71.5° (c=1 in water, 20h)
MDL No.: MFCD00067674
Formula:
C₆H₁₃NO₅ Melting Pt: ∼ 192 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00135831 CAS Number: 66-84-2 EINECS: 200-638-1 Merck Index: 14,4458 |
Specification Test Results
Optical rotation | +73° (c=1 in water, 20h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...