trans-3-Chloroacrylic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A15527-MD A15527-03 A15527-06
CAAAAA15527-MDEA 82.32 CAD
CAAAAA15527-MD CAAAA15527-03 CAAAA15527-06
trans-3-Chloroacrylic acid 99%
trans-3-Chloroacrylic acid

Purity: 99% CAS No.: 2345-61-1 Molecular Formula: C3H3ClO2 Formula Weight: 106.51 Storage Temperature: 2°C to 8°C Melting Point: 81-85° Beilstein Reference: 1720279 UN No.: 3261 MDL No.: MFCD00064237

Formula: C₃H₃ClO₂
MW: 106.51
Melting Pt: 81…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064237
CAS Number: 2345-61-1
EINECS: 219-070-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR