4-Chlorophenyl phosphorodichloridate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15507-14
A15507-22
CAAAA15507-14EA
153.73
CAD
CAAAA15507-14
CAAAA15507-22
4-Chlorophenyl phosphorodichloridate 98+%
4-Chlorophenyl phosphorodichloridate
Purity: 98+%
CAS No.: 772-79-2
Molecular Formula: C6H4Cl3O2P
Formula Weight: 245.43
Flash Point: 106°(222°F)
Refractive Index: 1.5398
Odor: Acrid
Boiling Point: 113-115°/0.9mm
Beilstein Reference: 910754
UN No.: 3265
MDL No.: MFCD00009705
Formula:
C₆H₄Cl₃O₂P Boiling Pt: 113…115 °C/0.9 mm Density: 1.508 Flash Pt: 106 °C(222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009705 CAS Number: 772-79-2 EINECS: 212-254-1 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...