3-(1-Naphthyl)acrylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Naphthalen-1-yl)acrylic acid
A15422-09
A15422-18
A15422-30
CAAAA15422-09EA
100.17
CAD
CAAAA15422-09
CAAAA15422-18
CAAAA15422-30
3-(1-Naphthyl)acrylic acid 98%
3-(1-Naphthyl)acrylic acid
Purity: 98%
CAS No.: 13026-12-5
Molecular Formula: C13H10O2
Formula Weight: 198.22
Melting Point: 210-212°
Beilstein Reference: 1868617
MDL No.: MFCD00014317
Formula:
C₁₃H₁₀O₂ MW: 198.22 g/mol Melting Pt: 210…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014317 CAS Number: 13026-12-5 EINECS: 235-887-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...