3,5-Dimethylisoxazole-4-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,5-Dimethylisoxasole-4-carboxylic acid

A15419-06 A15419-14
CAAAA15419-03EA 119.65 CAD
CAAAA15419-03 CAAAA15419-06 CAAAA15419-14
3,5-Dimethylisoxazole-4-carboxylic acid 99%
3,5-Dimethylisoxazole-4-carboxylic acid

Purity: 99% CAS No.: 2510-36-3 Molecular Formula: C6H7NO3 Formula Weight: 141.13 Melting Point: 140-142° Beilstein Reference: 115819

Formula: C₆H₇NO₃
MW: 141.13 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00051657
CAS Number: 2510-36-3
EINECS: 219-724-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR