3-Chloropropylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A15415-18 A15415-30
CAAAA15415-0BEA 775.76 CAD
CAAAA15415-0B CAAAA15415-18 CAAAA15415-30
3-Chloropropylamine hydrochloride 98%
3-Chloropropylamine hydrochloride

Purity: 98% CAS No.: 6276-54-6 Molecular Formula: Cl(CH2)3NH2.HCl Formula Weight: 130.02 Melting Point: 147-152° Beilstein Reference: 3667451 MDL No.: MFCD00012913

Formula: C₃H₈ClN
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00012913
CAS Number: 6276-54-6
EINECS: 228-474-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR