Terephthalaldehyde mono(diethyl acetal) 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15413-06
A15413-14
A15413-22
CAAAAA15413-06EA
109.18
CAD
CAAAAA15413-06
CAAAAA15413-36
CAAAA15413-14
CAAAA15413-22
Terephthalaldehyde mono(diethyl acetal) 98% stabilized
Terephthalaldehyde mono(diethyl acetal)
Purity: 98%
CAS No.: 81172-89-6
Molecular Formula: C12H16O3
Formula Weight: 208.26
Flash Point:
Formula:
C₁₂H₁₆O₃ Boiling Pt: 89…93 °C/7 mm Density: 1.047 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010217 CAS Number: 81172-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...