DL-2,4-Diaminobutyric acid dihydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-Dab·2HCl, H-DL-Dab.2HCl

A15408-03 A15408-06 A15408-14
CAAAA15408-03EA 144.41 CAD
CAAAA15408-03 CAAAA15408-06 CAAAA15408-14
DL-2,4-Diaminobutyric acid dihydrochloride 99%
DL-2,4-Diaminobutyric acid dihydrochloride

Purity: 99% CAS No.: 65427-54-5 Molecular Formula: C4H10N2O2.2HCl Formula Weight: 191.06 Melting Point: ca 198° dec. Beilstein Reference: 4708499 MDL No.: MFCD00012910

Formula: C4H12Cl2N2O2
MW: 191.1 g/mol
Melting Pt: ca 198 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012910
CAS Number: 65427-54-5
EINECS: 265-774-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR