Solvent Blue 38
Supplier: Thermo Scientific Chemicals
Synonyms:
Direct Blue 86, C.I. 74180, Luxol® Fast Blue MBSN
A15395-14
A15395-22
CAAAA15395-14EA
82.48
CAD
CAAAA15395-14
CAAAA15395-22
Solvent Blue 38
Solvent Blue 38
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.
CAS No.: 1328-51-4
Odor: Odorless
Melting Point: ca 235° dec.
MDL No.: MFCD00071424
Formula:
C32H12CuN8Na2O6S2 Melting Pt: ca 235 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00071424 CAS Number: 1328-51-4 EINECS: 215-523-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...