Esculetin ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15393-03
A15393-06
CAAAA15393-03EA
113.29
CAD
CAAAA15393-03
CAAAA15393-06
Esculetin ≥98%
Esculetin
A lipoxygenase inhibitor
Purity: 98+%
CAS No.: 305-01-1
Molecular Formula: C9H6O4
Formula Weight: 178.15
Storage Temperature: 2°C to 8°C
Melting Point: ca 270° dec.
Beilstein Reference: 152788
MDL No.: MFCD00006874
Formula:
C₉H₆O₄ Melting Pt: ∼ 270 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006874 CAS Number: 305-01-1 EINECS: 206-161-5 Merck Index: 14,3697 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...